Anti-TMPRSS4 Rabbit Polyclonal Antibody
AVIVARP49463_P050
- Antibody type:Primary
- Antigen name:transmembrane protease, serine 4
- Antigen symbol:TMPRSS4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:56649
- Antigen synonyms:CAPH2|TMPRSS3|MT-SP2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: TMPRSS4 (transmembrane protease, serine 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: