Anti-UGT1A5 Rabbit Polyclonal Antibody
Catalog # AVIVARP49439_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:UDP glucuronosyltransferase 1 family, polypeptide A5
- Antigen symbol:UGT1A5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:54579
- Antigen synonyms:UDPGT 1-5|UGT1E|UDPGT
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: UGT1A5 (UDP glucuronosyltransferase 1 family, polypeptide A5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human