Anti-MTFMT Rabbit Polyclonal Antibody
AVIVARP49013_P050
- Antibody type:Primary
- Antigen name:mitochondrial methionyl-tRNA formyltransferase
- Antigen symbol:MTFMT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Pig
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:123263
- Antigen synonyms:COXPD15|FMT1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: MTFMT (mitochondrial methionyl-tRNA formyltransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Pig