Order Entry
Export
ContactUsLinkComponent
Anti-POLR3A Rabbit Polyclonal Antibody
Anti-POLR3A Rabbit Polyclonal Antibody
Catalog # AVIVARP48725_P050
undefined
Anti-POLR3A Rabbit Polyclonal Antibody
Catalog # AVIVARP48725_P050

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Polymerase (RNA) III (DNA directed) polypeptide A, 155kDa
  • Antigen symbol:
    POLR3A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    middle
  • Cross adsorption:
    none
  • Gene ID:
    11128
  • Antigen synonyms:
    HLD7|ADDH|RPC155|hRPC155|RPC1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: POLR3A (polymerase (RNA) III (DNA directed) polypeptide A, 155kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: