Anti-ZNF256 Rabbit Polyclonal Antibody
AVIVARP47598_P050
- Antibody type:Primary
- Antigen name:zinc finger protein 256
- Antigen symbol:ZNF256
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:10172
- Antigen synonyms:BMZF-3|BMZF3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: ZNF256 (zinc finger protein 256)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human