Anti-ZKSCAN1 Rabbit Polyclonal Antibody
Catalog # AVIVARP47580_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:zinc finger with KRAB and SCAN domains 1
- Antigen symbol:ZKSCAN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:7586
- Antigen synonyms:ZNF36|ZSCAN33|9130423L19Rik|PHZ-37|ZNF139|KOX18
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: ZKSCAN1 (zinc finger with KRAB and SCAN domains 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: