Anti-CTH Rabbit Polyclonal Antibody
AVIVARP46068_T100
- Antibody type:Primary
- Antigen name:cystathionine gamma-lyase
- Antigen symbol:CTH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:1491
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Type: Primary
Antigen: CTH (cystathionase (cystathionine gamma-lyase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: