Anti-BLK Rabbit Polyclonal Antibody
Catalog # AVIVARP46061_T100
Supplier: Aviva Systems Biology
Jump to:
Product Details & Documents
Type: Primary
Antigen: BLK (B lymphoid tyrosine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Documents
ARP46061_T100
Datasheet
Specifications
- Western blot:Yes
- Clonality:Polyclonal
- Antibody type:Primary
- Epitope:middle
- Storage temperature:Cool
- Host:Rabbit
- Cross adsorption:none
- Shipping temperature:Ice
- Antigen name:BLK proto-oncogene, Src family tyrosine kinase
- Cat. No.:AVIVARP46061_T100
- Gene ID:640
- Antigen synonyms:MODY11
- Conjugation:Unconjugated
- Antigen symbol:BLK



