Anti-EIF2S3 Rabbit Polyclonal Antibody
Catalog # AVIVARP46056_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Eukaryotic translation initiation factor 2, subunit 3 gamma, 52kDa
- Antigen symbol:EIF2S3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:1968
- Antigen synonyms:EIF2gamma|EIF2|EIF2G|eIF-2gA
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: EIF2S3 (eukaryotic translation initiation factor 2, subunit 3 gamma, 52kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: