Anti-SKAP1 Rabbit Polyclonal Antibody
Catalog # AVIVARP46047_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Src kinase associated phosphoprotein 1
- Antigen symbol:SKAP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:8631
- Antigen synonyms:SKAP55|HEL-S-81p|SCAP1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: SKAP1 (src kinase associated phosphoprotein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: