Anti-RORA Rabbit Polyclonal Antibody
Catalog # AVIVARP45607_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:RAR-related orphan receptor A
- Antigen symbol:RORA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:6095
- Antigen synonyms:RZRA|RZR-ALPHA|ROR3|ROR1|NR1F1|ROR2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: RORA (RAR-related orphan receptor A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: