Anti-YIF1B Rabbit Polyclonal Antibody
Catalog # AVIVARP44885_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Yip1 interacting factor homolog B (S. cerevisiae)
- Antigen symbol:YIF1B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:90522
- Antigen synonyms:FinGER8
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: YIF1B (Yip1 interacting factor homolog B (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: