Anti-ZDHHC19 Rabbit Polyclonal Antibody
Catalog # AVIVARP44883_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:zinc finger, DHHC-type containing 19
- Antigen symbol:ZDHHC19
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:131540
- Antigen synonyms:DHHC19
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: ZDHHC19 (zinc finger, DHHC-type containing 19)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human