Anti-SLC6A5 Rabbit Polyclonal Antibody
Catalog # AVIVARP43872_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 6 (neurotransmitter transporter), member 5
- Antigen symbol:Solute carrier family 6, member 5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: SLC6A5 (solute carrier family 6 (neurotransmitter transporter), member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human