Anti-SLC30A3 Rabbit Polyclonal Antibody
AVIVARP43848_P050
- Antibody type:Primary
- Antigen name:solute carrier family 30 (zinc transporter), member 3
- Antigen symbol:Solute carrier family 30, member 3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:7781
- Antigen synonyms:ZNT3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: SLC30A3 (solute carrier family 30 (zinc transporter), member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: