Anti-ABCA5 Rabbit Polyclonal Antibody
Catalog # AVIVARP43688_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:ATP-binding cassette, sub-family A (ABC1), member 5
- Antigen symbol:ABCA5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:23461
- Antigen synonyms:ABC13|EST90625
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: ABCA5 (ATP-binding cassette, sub-family A (ABC1), member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: