Anti-NIPSNAP3A Rabbit Polyclonal Antibody
Catalog # AVIVARP43680_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:nipsnap homolog 3A (C. elegans)
- Antigen symbol:NIPSNAP3A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:25934
- Antigen synonyms:HSPC299|TASSC|NIPSNAP4
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: NIPSNAP3A (nipsnap homolog 3A (C. elegans))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: