Anti-ABCF2 Rabbit Polyclonal Antibody
Catalog # AVIVARP43666_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:ATP-binding cassette, sub-family F (GCN20), member 2
- Antigen symbol:ABCF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:10061
- Antigen synonyms:HUSSY18|EST133090|ABC28|HUSSY-18
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: ABCF2 (ATP-binding cassette, sub-family F (GCN20), member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: