Anti-ABCC9 Rabbit Polyclonal Antibody
Catalog # AVIVARP43664_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:ATP-binding cassette, sub-family C (CFTR/MRP), member 9
- Antigen symbol:ABCC9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:10060
- Antigen synonyms:SUR2|CANTU|ATFB12|CMD1O|ABC37
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: ABCC9 (ATP-binding cassette, sub-family C (CFTR/MRP), member 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: