Anti-ATAT1 Rabbit Polyclonal Antibody
Catalog # AVIVARP42643_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Alpha tubulin acetyltransferase 1
- Antigen symbol:ATAT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Pig,Human,Bovine,Dog,Rat,Mouse
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:79969
- Antigen synonyms:MEC17|Nbla00487|TAT|C6orf134
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: ATAT1 (alpha tubulin acetyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Mouse, Pig, Rat