Anti-GPD1 Rabbit Polyclonal Antibody
Catalog # AVIVARP42227_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Glycerol-3-phosphate dehydrogenase 1 (soluble)
- Antigen symbol:GPD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:2819
- Antigen synonyms:GPD-C|HTGTI|GPDH-C
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: GPD1 (glycerol-3-phosphate dehydrogenase 1 (soluble))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: