Anti-CD8B Rabbit Polyclonal Antibody
Catalog # AVIVARP42204_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:CD8b molecule
- Antigen symbol:CD8B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:926
- Antigen synonyms:LY3|LYT3|LEU2|P37|CD8B1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: CD8B (CD8b molecule)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: