Order Entry
Export
ContactUsLinkComponent
 
Anti-MAP2K3 Rabbit Polyclonal Antibody
  AVIVARP42065_P050
Anti-MAP2K3 Rabbit Polyclonal Antibody
  AVIVARP42065_P050

 

  • Antibody type:
    Primary
  • Antigen name:
    Mitogen-activated protein kinase kinase 3
  • Antigen symbol:
    MAP2K3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Cross adsorption:
    none
  • Gene ID:
    5606
  • Antigen synonyms:
    SAPKK-2|MAPKK3|MEK3|SAPKK2|PRKMK3|MKK3
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: MAP2K3 (mitogen-activated protein kinase kinase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: