Anti-MMP1 Rabbit Polyclonal Antibody
Catalog # AVIVARP42039_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:matrix metallopeptidase 1
- Antigen symbol:MMP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:4312
- Antigen synonyms:CLGN|CLG
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: MMP1 (matrix metallopeptidase 1 (interstitial collagenase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: