Anti-LAIR2 Rabbit Polyclonal Antibody
AVIVARP42032_P050
- Antibody type:Primary
- Antigen name:leukocyte-associated immunoglobulin-like receptor 2
- Antigen symbol:LAIR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:3904
- Antigen synonyms:CD306
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: LAIR2 (leukocyte-associated immunoglobulin-like receptor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human