Anti-CEACAM6 Rabbit Polyclonal Antibody
Catalog # AVIVARP41504_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Carcinoembryonic antigen-related cell adhesion molecule 6
- Antigen symbol:CEACAM6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:4680
- Antigen synonyms:CEAL|NCA|CD66c
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: CEACAM6 (carcinoembryonic antigen-related cell adhesion molecule 6 (non-specific cross reacting antigen))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human