Order Entry
Export
ContactUsLinkComponent
 
Anti-DDC Rabbit Polyclonal Antibody
Catalog # AVIVARP41425_T100
Anti-DDC Rabbit Polyclonal Antibody
Catalog # AVIVARP41425_T100

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    DOPA Decarboxylase
  • Antigen symbol:
    DDC
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Cross adsorption:
    none
  • Gene ID:
    1644
  • Antigen synonyms:
    AADC
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: DDC (dopa decarboxylase (aromatic L-amino acid decarboxylase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: