Order Entry
Export
ContactUsLinkComponent
Anti-SFTPB Rabbit Polyclonal Antibody
Anti-SFTPB Rabbit Polyclonal Antibody
  AVIVARP41411_P050
Anti-SFTPB Rabbit Polyclonal Antibody
  AVIVARP41411_P050

 

  • Antibody type:
    Primary
  • Antigen name:
    Surfactant Protein B
  • Antigen symbol:
    SFTPB
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Pig,
    Human,
    Guinea pig,
    Dog,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    middle
  • Cross adsorption:
    none
  • Gene ID:
    6439
  • Antigen synonyms:
    SMDP1|PSP-B|SP-B|SFTP3|SFTB3
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: SFTPB (surfactant protein B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Dog, Guinea pig, Human, Mouse, Pig, Rat