Anti-STAU1 Rabbit Polyclonal Antibody
AVIVARP40536_P050
- Antibody type:Primary
- Antigen name:Staufen double-stranded RNA binding protein 1
- Antigen symbol:STAU1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:6780
- Antigen synonyms:PPP1R150|STAU
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: STAU1 (staufen double-stranded RNA binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: