Anti-NOL4 Rabbit Polyclonal Antibody
AVIVARP40492_P050
- Antibody type:Primary
- Antigen name:Nucleolar protein 4
- Antigen symbol:NOL4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:8715
- Antigen synonyms:CT125|HRIHFB2255|NOLP
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: NOL4 (nucleolar protein 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: