Anti-RNASE1 Rabbit Polyclonal Antibody
Catalog # AVIVARP40432_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Ribonuclease, RNase A family, 1 (pancreatic)
- Antigen symbol:RNASE1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:6035
- Antigen synonyms:RIB1|RAC1|RNS1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: RNASE1 (ribonuclease, RNase A family, 1 (pancreatic))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat