Anti-NOVA2 Rabbit Polyclonal Antibody
Catalog # AVIVARP40399_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:neuro-oncological ventral antigen 2
- Antigen symbol:NOVA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:4858
- Antigen synonyms:ANOVA|NOVA3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: NOVA2 (neuro-oncological ventral antigen 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: