Anti-Zfp74 Rabbit Polyclonal Antibody
Catalog # AVIVARP39903_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:zinc finger protein 74
- Antigen symbol:ZFP74
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:72723
- Antigen synonyms:Zfp66|mszf77|Znf569|KRAB8|mszf21|2810054M15Rik
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: Zfp74
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: