Anti-ZNF496 Rabbit Polyclonal Antibody
Catalog # AVIVARP39715_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:zinc finger protein 496
- Antigen symbol:ZNF496
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:84838
- Antigen synonyms:ZKSCAN17|ZFP496|ZSCAN49|NIZP1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: ZNF496 (zinc finger protein 496)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: