Anti-TRIM8 Rabbit Polyclonal Antibody
Catalog # AVIVARP39655_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:tripartite motif containing 8
- Antigen symbol:TRIM8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:81603
- Antigen synonyms:GERP|RNF27
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: TRIM8 (tripartite motif containing 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: