Anti-NFYC Rabbit Polyclonal Antibody
Catalog # AVIVARP39066_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Nuclear Transcription Factor Y, gamma
- Antigen symbol:NFYC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:4802
- Antigen synonyms:HAP5|NF-YC|H1TF2A|CBFC|HSM|CBF-C
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: NFYC (nuclear transcription factor Y, gamma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: