Anti-POU6F2 Rabbit Polyclonal Antibody
AVIVARP38929_P050
- Antibody type:Primary
- Antigen name:POU class 6 homeobox 2
- Antigen symbol:POU6F2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:11281
- Antigen synonyms:RPF-1|WT5|WTSL
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: POU6F2 (POU class 6 homeobox 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: