Order Entry
Export
ContactUsLinkComponent
Anti-NR4A2 Rabbit Polyclonal Antibody
Anti-NR4A2 Rabbit Polyclonal Antibody
Catalog # AVIVARP38754_P050
Anti-NR4A2 Rabbit Polyclonal Antibody
Catalog # AVIVARP38754_P050

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    nuclear receptor subfamily 4, group A, member 2
  • Antigen symbol:
    NR4A2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    middle
  • Cross adsorption:
    none
  • Gene ID:
    4929
  • Antigen synonyms:
    NOT|RNR1|NURR1|HZF-3|TINUR
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: NR4A2 (nuclear receptor subfamily 4, group A, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: