Anti-TCF15 Rabbit Polyclonal Antibody
Catalog # AVIVARP38503_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:transcription factor 15 (basic helix-loop-helix)
- Antigen symbol:TCF15
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:6939
- Antigen synonyms:bHLHa40|PARAXIS|EC2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: TCF15 (transcription factor 15 (basic helix-loop-helix))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: