Anti-NFATC4 Rabbit Polyclonal Antibody
Catalog # AVIVARP38493_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 4
- Antigen symbol:NFATC4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:4776
- Antigen synonyms:NF-AT3|NFAT3|NF-ATC4
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: NFATC4 (nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: