Anti-HOXB7 Rabbit Polyclonal Antibody
Catalog # AVIVARP38482_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:homeobox B7
- Antigen symbol:HOXB7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:3217
- Antigen synonyms:HOX2C|Hox-2.3|HOX2|HHO.C1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: HOXB7 (homeobox B7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: