Anti-ONECUT1 Rabbit Polyclonal Antibody
AVIVARP38480_P050
- Antibody type:Primary
- Antigen name:one cut homeobox 1
- Antigen symbol:ONECUT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:3175
- Antigen synonyms:HNF6A|HNF-6|HNF6
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: ONECUT1 (one cut homeobox 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: