Anti-TRIP13 Rabbit Polyclonal Antibody
Catalog # AVIVARP38434_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Thyroid Hormone Receptor Interactor 13
- Antigen symbol:TRIP13
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:9319
- Antigen synonyms:16E1BP
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: TRIP13 (thyroid hormone receptor interactor 13)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: