Anti-TANK Rabbit Polyclonal Antibody
AVIVARP38420_P050
- Antibody type:Primary
- Antigen name:TRAF family member-associated NFKB activator
- Antigen symbol:TANK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:10010
- Antigen synonyms:ITRAF|I-TRAF|TRAF2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: TANK (TRAF family member-associated NFKB activator)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: