Anti-TFAP2B Rabbit Polyclonal Antibody
Catalog # AVIVARP38283_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Transcription Factor AP-2 beta
- Antigen symbol:TFAP2B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:7021
- Antigen synonyms:AP-2B|AP2-B
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: TFAP2B (transcription factor AP-2 beta (activating enhancer binding protein 2 beta))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: