Anti-STAT1 Rabbit Polyclonal Antibody
Catalog # AVIVARP36979_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Signal Transducer And Activator of Transcription 1
- Antigen symbol:STAT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:20846
- Antigen synonyms:AA408197|2010005J02Rik
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: STAT1 (signal transducer and activator of transcription 1, 91kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: