Anti-DDX27 Rabbit Polyclonal Antibody
Catalog # AVIVARP36447_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:DEAD (Asp-Glu-Ala-Asp) box polypeptide 27
- Antigen symbol:DDX27
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:55661
- Antigen synonyms:PP3241|DRS1|RHLP|dJ686N3.1|Drs1p|HSPC259
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: DDX27 (DEAD (Asp-Glu-Ala-Asp) box polypeptide 27)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: