Anti-ZCCHC17 Rabbit Polyclonal Antibody
Catalog # AVIVARP36444_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:zinc finger, CCHC domain containing 17
- Antigen symbol:ZCCHC17
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:51538
- Antigen synonyms:HSPC251|pNO40|PS1D
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: ZCCHC17 (zinc finger, CCHC domain containing 17)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: