Anti-ZNF75D Rabbit Polyclonal Antibody
AVIVARP35823_T100
- Antibody type:Primary
- Antigen name:zinc finger protein 75D
- Antigen symbol:ZNF75D
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:7626
- Antigen synonyms:ZNF75|ZNF82|ZSCAN28|D8C6|ZKSCAN24
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Type: Primary
Antigen: ZNF75D (zinc finger protein 75D)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: