Anti-CLIC2 Rabbit Polyclonal Antibody
Catalog # AVIVARP35046_T100
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:chloride intracellular channel 2
- Antigen symbol:CLIC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Pig,Rabbit,Human,Bovine,Dog,Rat
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:1193
- Antigen synonyms:MRXS32|CLIC2b|XAP121
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: CLIC2 (chloride intracellular channel 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Pig, Rabbit, Rat